Learn More
Abnova™ Human PANK2 Partial ORF (NP_705902.2, 437 a.a. - 570 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a protein belonging to the pantothenate kinase family and is the only member of that family to be expressed in mitochondria. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by acyl CoA species. Mutations in this gene are associated with HARP syndrome and pantothenate kinase-associated neurodegeneration (PKAN), formerly Hallervorden-Spatz syndrome. Alternative splicing, involving the use of alternate first exons, results in multiple transcripts encoding different isoforms. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_705902.2 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80025 |
Molecular Weight (g/mol) | 41.2kDa |
Name | PANK2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 25 μg |
Immunogen | MASRGDSTKVDKLVRDIYGGDYERFGLPGWAVASSFGNMMSKEKREAVSKEDLARATLITITNNIGSIARMCALNENINQVVFVGNFLRINTIAMRLLAYALDYWSKGQLKALFSEHEGYFGAVGALLELLKIP |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.