Learn More
Abnova™ Human PAPSS1 Partial ORF (NP_005434, 144 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 [PubMed 10679223]). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2 (MIM 603005).[supplied by OMIM]
Specifications
Specifications
Accession Number | NP_005434 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9061 |
Molecular Weight (g/mol) | 37.73kDa |
Name | PAPSS1 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | QIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAE |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.