missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PBX4 Partial ORF (NP_079521, 319 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16125003
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a homeodomain protein with similarity to a transcription factor involved in translocations in pre-B-cell leukemias. The presence of a homeobox domain suggests that this protein acts as a transcription factor, but this function has not been demonstrated. [provided by RefSeq]

Sequence: AGDAFLTLRTLASLQPPPGGGCLQSQAQGSWQGATPQPATASPAGDPGSINSSTSN

Specifications

Accession Number NP_079521
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80714
Molecular Weight (g/mol) 31.9kDa
Name PBX4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen AGDAFLTLRTLASLQPPPGGGCLQSQAQGSWQGATPQPATASPAGDPGSINSSTSN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name PBX4
Gene Symbol PBX4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human PBX4 Partial ORF (NP_079521, 319 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.