missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human PCDHB15 Full-length ORF (AAH38797, 1 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16162073

Abnova™ Human PCDHB15 Full-length ORF (AAH38797, 1 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16162073
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16162073

Brand: Abnova™ H00056121P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: MEPAGERFPEQRQVLILLLLLEVTLAGWEPRRYSVMEETERGSFVANLANDLGLGVGELAERGARVVSEDNEQGLQLDLQTGQLILNEKLDREKLCGPTEPCIMHFQVLLKKPLEVFRAELLVTDINDHSPEFPEREMTLKIPETSSLGTVFPLKKARDLDVGSNNVQNYNISPNSHFHVSTRTRGDGRKYPELVLDTELDREEQAELRLTLTAVDGGSPPRSGTVQILILVLDANDNAPEFVQALYEVQVPENSPVGSLVVKVSARDLDTGTNGEISYSLYYSSQEIDKPFELSSLSGEIRLIKKLDFETMSSYDLDIEASDGGGLSGKCSVSVKVLDVNDNFPELSISSLTSPIPENSPETEVALFRIRDRDSGENGKMICSIQDDVPFKLKPSVENFYRLVTEGALDRETRAEYNITITITDLGTPRLKTEQSITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVRATDRDSGTNAQVTYSLLPPQDPHLPLTSLVSINTDNGHLFALQSLDYEALQAFEFRVGATDRGFPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDVAKHRLVVLVKDNGEPPRSATATLQVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVFLFVAVRLCRRSRAASVGRCSVPEGPFPGHLVDVSGTGTLSQSYQYEVCLTGGSESNDFKFLKPIFPNIVSQDSRRKSEFLE

Specifications

Accession Number AAH38797
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56121
Molecular Weight (g/mol) 112.31kDa
Name PCDHB15 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEPAGERFPEQRQVLILLLLLEVTLAGWEPRRYSVMEETERGSFVANLANDLGLGVGELAERGARVVSEDNEQGLQLDLQTGQLILNEKLDREKLCGPTEPCIMHFQVLLKKPLEVFRAELLVTDINDHSPEFPEREMTLKIPETSSLGTVFPLKKARDLDVGSNNVQNYNISPNSHFHVSTRTRGDGRKYPELVLDTELDREEQAELRLTLTAVDGGSPPRSGTVQILILVLDANDNAPEFVQALYEVQVPENSPVGSLVVKVSARDLDTGTNGEISYSLYYSSQEIDKPFELSSLSGEIRLIKKLDFETMSSYDLDIEASDGGGLSGKCSVSVKVLDVNDNFPELSISSLTSPIPENSPETEVALFRIRDRDSGENGKMICSIQDDVPFKLKPSVENFYRLVTEGALDRETRAEYNITITITDLGTPRLKTEQSITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVRATDRDSGTNAQVTYSLLPPQDPHLPLTSLVSINTDNGHLFALQSLDYEALQAFEFRVGATDRGFPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDVAKHRLVVLVKDNGEPPRSATATLQVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVFLFVAVRLCRRSRAASVGRCSVPEGPFPGHLVDVSGTGTLSQSYQYEVCLTGGSESNDFKFLKPIFPNIVSQDSRRKSEFLE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PCDH-BETA15
Common Name PCDHB15
Gene Symbol PCDHB15
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human PCDHB15 Full-length ORF (AAH38797, 1 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.