Learn More
Abnova™ Human PER3 Full-length ORF (AAH26102.1, 1 a.a. - 378 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008863-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq]
Sequence: MPRGEAPGPGRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQILSQNGAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAALILNRKKDVLASSHFVDLLAPQDMRVFYAHTARAQLPFWNNWTQRAARYECAPVKPFFCRIRGGEDRKQEKCHSPFRIIPYLIHVHHPAQPELESEPCCLTVVEKIHSGYEAPRIPVNKRIFTTTHTPGCVFLEVDEKAVPLLGYLPQDLIGTSILSYLHPEDRSLMVAIHQKVLKYAGHPPFEHSPIRFCTQNGDYIILDSSWSSFVNPWSRKISFIIGRHKVRTSpecifications
AAH26102.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GIG13 | |
PER3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8863 | |
PER3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPRGEAPGPGRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQILSQNGAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAALILNRKKDVLASSHFVDLLAPQDMRVFYAHTARAQLPFWNNWTQRAARYECAPVKPFFCRIRGGEDRKQEKCHSPFRIIPYLIHVHHPAQPELESEPCCLTVVEKIHSGYEAPRIPVNKRIFTTTHTPGCVFLEVDEKAVPLLGYLPQDLIGTSILSYLHPEDRSLMVAIHQKVLKYAGHPPFEHSPIRFCTQNGDYIILDSSWSSFVNPWSRKISFIIGRHKVRT | |
RUO | |
PER3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |