Learn More
Abnova™ Human PER3 Full-length ORF (AAH26102.1, 1 a.a. - 378 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH26102.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8863 |
Molecular Weight (g/mol) | 69.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173012
|
Abnova™
H00008863-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16163012
|
Abnova™
H00008863-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq]
Sequence: MPRGEAPGPGRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQILSQNGAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAALILNRKKDVLASSHFVDLLAPQDMRVFYAHTARAQLPFWNNWTQRAARYECAPVKPFFCRIRGGEDRKQEKCHSPFRIIPYLIHVHHPAQPELESEPCCLTVVEKIHSGYEAPRIPVNKRIFTTTHTPGCVFLEVDEKAVPLLGYLPQDLIGTSILSYLHPEDRSLMVAIHQKVLKYAGHPPFEHSPIRFCTQNGDYIILDSSWSSFVNPWSRKISFIIGRHKVRTSpecifications
AAH26102.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPRGEAPGPGRRGAKDEALGEESGERWSPEFHLQRKLADSSHSEQQDRNRVSEELIMVVQEMKKYFPSERRNKPSTLDALNYALRCVHSVQANSEFFQILSQNGAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAALILNRKKDVLASSHFVDLLAPQDMRVFYAHTARAQLPFWNNWTQRAARYECAPVKPFFCRIRGGEDRKQEKCHSPFRIIPYLIHVHHPAQPELESEPCCLTVVEKIHSGYEAPRIPVNKRIFTTTHTPGCVFLEVDEKAVPLLGYLPQDLIGTSILSYLHPEDRSLMVAIHQKVLKYAGHPPFEHSPIRFCTQNGDYIILDSSWSSFVNPWSRKISFIIGRHKVRT | |
RUO | |
PER3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8863 | |
PER3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GIG13 | |
PER3 | |
Recombinant | |
wheat germ expression system |