missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human PER3 Partial ORF (NP_058515.1, 1105 a.a. - 1201 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16179485

Abnova™ Human PER3 Partial ORF (NP_058515.1, 1105 a.a. - 1201 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16179485
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16179485

Brand: Abnova™ H00008863Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq]

Sequence: WRMIRQTPERILMTYQVPERVKEVVLKEDLEKLESMRQQQPQFSHGQKEELAKVYNWIQSQTVTQEIDIQACVTCENEDSADGAATSCGQVLVEDSC

Specifications

Accession Number NP_058515.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8863
Molecular Weight (g/mol) 36.41kDa
Name PER3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen WRMIRQTPERILMTYQVPERVKEVVLKEDLEKLESMRQQQPQFSHGQKEELAKVYNWIQSQTVTQEIDIQACVTCENEDSADGAATSCGQVLVEDSC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GIG13
Common Name PER3
Gene Symbol PER3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human PER3 Partial ORF (NP_058515.1, 1105 a.a. - 1201 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.