missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-DDR1/DDR2 (DDR1 Y796, DDR2 Y740) Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB25382
This item is not returnable.
View return policy
Description
DDR1/DDR2 Monoclonal specifically detects DDR1/DDR2 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
DDR2 | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 ÎĽm filtered solution in PBS. with Sodium Azide | |
CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKT, TKTMIG20a, Trk3, Tyro-10, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
4921 | |
Human | |
Supernatant |
Western Blot | |
1119D | |
Western Blot 1:1000 dilution, Simple Western 1:100 dilution | |
Q16832 | |
DDR2 | |
Phosphopeptide containing the human DDR2 Y740 site Accession # Q16832 | |
100 ÎĽg | |
Primary | |
Detects human DDR2 when phosphorylated at Y740. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Phospho-DDR1/DDR2 (DDR1 Y796, DDR2 Y740) Antibody, R&D Systems™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction