Learn More
Abnova™ Human PIGF Partial ORF (NP_775097.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005281-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor, a glycolipid containing three mannose molecules in its core backbone, is found on many blood cells where it serves to anchor proteins to the cell surface. The encoded protein and another GPI synthesis protein, PIGO, function in the transfer of ethanolaminephosphate to the third mannose in GPI. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGFVTAVNLVLYLVVKPNTSSKRSSLSHKVTGFLKCCIYFLMSCFSFHVIFVLSpecifications
NP_775097.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGFVTAVNLVLYLVVKPNTSSKRSSLSHKVTGFLKCCIYFLMSCFSFHVIFVL | |
RUO | |
PIGF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5281 | |
PIGF (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC32646/MGC33136 | |
PIGF | |
Recombinant | |
wheat germ expression system |