missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human PIK4CA Partial ORF (NP_002641, 338 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16132835

Abnova™ Human PIK4CA Partial ORF (NP_002641, 338 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16132835
10 ug, 10µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16132835

Brand: Abnova™ H00005297Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: VPEAIKFLVTWHTIDADAPELSHVLCWAPTDPPTGLSYFSSMYPPHPLTAQYGVKVLRSFPPDAILFYIPQIVQALRYDKMGYVREYILWAASKSQLLAHQFIWNMKTNI

Specifications

Accession Number NP_002641
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5297
Molecular Weight (g/mol) 37.84kDa
Name PIK4CA (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen VPEAIKFLVTWHTIDADAPELSHVLCWAPTDPPTGLSYFSSMYPPHPLTAQYGVKVLRSFPPDAILFYIPQIVQALRYDKMGYVREYILWAASKSQLLAHQFIWNMKTNI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ16556/PI4K-ALPHA/PIK4CA/pi4K230
Common Name PI4KA
Gene Symbol PI4KA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human PIK4CA Partial ORF (NP_002641, 338 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.