missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PODN Partial ORF (NP_714914.2, 541 a.a. - 644 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00127435-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: GPRAWVDLAHLQLLDIAGNQLTEIPEGLPESLEYLYLQNNKISAVPANAFDSTPNLKGIFLRFNKLAVGSVVDSAFRRLKHLQVLDIEGNLEFGDISKDRGRLGSpecifications
NP_714914.2 | |
Liquid | |
127435 | |
PODN (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC24995/PCAN/SLRR5A | |
PODN | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.18kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GPRAWVDLAHLQLLDIAGNQLTEIPEGLPESLEYLYLQNNKISAVPANAFDSTPNLKGIFLRFNKLAVGSVVDSAFRRLKHLQVLDIEGNLEFGDISKDRGRLG | |
RUO | |
PODN | |
Wheat Germ (in vitro) | |
GST |