Learn More
Abnova™ Human POLR3D Partial ORF (-, 296 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000661-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene complements a temperature-sensitive mutant isolated from the BHK-21 Syrian hamster cell line. It leads to a block in progression through the G1 phase of the cell cycle at nonpermissive temperatures. [provided by RefSeq]
Sequence: GQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDHSpecifications
Antibody Production, ELISA, Protein Array, Western Blot | |
661 | |
POLR3D (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BN51T/RPC4/RPC53/TSBN51 | |
POLR3D | |
Recombinant | |
wheat germ expression system |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDH | |
RUO | |
POLR3D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |