missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human POLR3D Partial ORF (-, 296 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
---|---|
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 661 |
Molecular Weight (g/mol) | 36.63kDa |
Name | POLR3D (Human) Recombinant Protein (Q01) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16192311
|
Abnova™
H00000661-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182311
|
Abnova™
H00000661-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene complements a temperature-sensitive mutant isolated from the BHK-21 Syrian hamster cell line. It leads to a block in progression through the G1 phase of the cell cycle at nonpermissive temperatures. [provided by RefSeq]
Sequence: GQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDHSpecifications
Antibody Production, ELISA, Protein Array, Western Blot | |
661 | |
POLR3D (Human) Recombinant Protein (Q01) | |
GQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDH | |
RUO | |
POLR3D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BN51T/RPC4/RPC53/TSBN51 | |
POLR3D | |
Recombinant | |
wheat germ expression system |