missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human POLR3E Full-length ORF (AAH00285, 1 a.a. - 666 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16181713

Abnova™ Human POLR3E Full-length ORF (AAH00285, 1 a.a. - 666 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16181713
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16181713

Brand: Abnova™ H00055718P01.10ug

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for AP, Array, ELISA, WB-Re

Sequence: MANEEDDPVVQEIDVYLAKSLAEKLYLFQYPVRPASMTYDDIPHLSAKIKPKQQKVELEMAIDTLNPNYCRSKGEQIALNVDGACADETSTYSSKLMDKQTFCSSQTTSNTSRYAAALYRQGELHLTPLHGILQLRPSFSYLDKADAKHREREAANEAGDSSQDEAEDDVKQITVRFSRPESEQARQRRVQSYEFLQKKHAEEPWVHLHYYGLRDSRSEHERQYLLCPGSSGVENTELVKSPSEYLMMLMPPSQEEEKDKPVAPSNVLSMAQLRTLPLADQIKILMKNVKVMPFANLMSLLGPSIDSVAVLRGIQKVAMLVQGNWVVKSDILYPKDSSSPHSGVPAEVLCRGRDFVMWKFTQSRWVVRKEVATVTKLCAEDVMDFLEHMAVVRINKGWEFILPYDGEFIKKHPDVVQRQHMLWTGIQAKLEKVYNLVKETMPKKPDAQSGPAGLVCGDQRIQVAKTKAQQNHALLERELQRRKEQLRVPAVPPGVRIKEEPVSEEGEEDEEQEAEEEPMDTSPSGLHSKLANGLPLGRAAGTDSFNGHPPQGCASTPVARELKAFVEATFQRQFVLTLSELKRLFNLHLASLPPGHTLFSGISDRMLQDTVLAAGCKQILVPFPPQTAASPDEQKVFALWESGDMSDQDCCVSYGGMWYLKGTVQS

Spezifikation

Accession Number AAH00285
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55718
Molecular Weight (g/mol) 99kDa
Name POLR3E (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MANEEDDPVVQEIDVYLAKSLAEKLYLFQYPVRPASMTYDDIPHLSAKIKPKQQKVELEMAIDTLNPNYCRSKGEQIALNVDGACADETSTYSSKLMDKQTFCSSQTTSNTSRYAAALYRQGELHLTPLHGILQLRPSFSYLDKADAKHREREAANEAGDSSQDEAEDDVKQITVRFSRPESEQARQRRVQSYEFLQKKHAEEPWVHLHYYGLRDSRSEHERQYLLCPGSSGVENTELVKSPSEYLMMLMPPSQEEEKDKPVAPSNVLSMAQLRTLPLADQIKILMKNVKVMPFANLMSLLGPSIDSVAVLRGIQKVAMLVQGNWVVKSDILYPKDSSSPHSGVPAEVLCRGRDFVMWKFTQSRWVVRKEVATVTKLCAEDVMDFLEHMAVVRINKGWEFILPYDGEFIKKHPDVVQRQHMLWTGIQAKLEKVYNLVKETMPKKPDAQSGPAGLVCGDQRIQVAKTKAQQNHALLERELQRRKEQLRVPAVPPGVRIKEEPVSEEGEEDEEQEAEEEPMDTSPSGLHSKLANGLPLGRAAGTDSFNGHPPQGCASTPVARELKAFVEATFQRQFVLTLSELKRLFNLHLASLPPGHTLFSGISDRMLQDTVLAAGCKQILVPFPPQTAASPDEQKVFALWESGDMSDQDCCVSYGGMWYLKGTVQS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias RPC5/SIN
Common Name POLR3E
Gene Symbol POLR3E
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Abnova™ Human POLR3E Full-length ORF (AAH00285, 1 a.a. - 666 a.a.) Recombinant Protein with GST-tag at N-terminal >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt