Learn More
Abnova™ Human POPDC2 Full-length ORF (NP_071418.2, 1 a.a. - 364 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064091-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the POP family of proteins which contain three putative transmembrane domains. This membrane associated protein is predominantly expressed in skeletal and cardiac muscle, and may have an important function in these tissues. [provided by RefSeq]
Sequence: MSANSSRVGQLLLQGSACIRWKQDVEGAVYHLANCLLLLGFMGGSGVYGCFYLFGFLSAGYLCCVLWGWFSACGLDIVLWSFLLAVVCLLQLAHLVYRLREDTLPEEFDLLYKTLCLPLQVPLQTYKEIVHCCEEQVLTLATEQTYAVEGETPINRLSLLLSGRVRVSQDGQFLHYIFPYQFMDSPEWESLQPSEEGVFQVTLTAETSCSYISWPRKSLHLLLTKERYISCLFSALLGYDISEKLYTLNDKLFAKFGLRFDIRLPSLYHVLGPTAADAGPESEKGDEEVCEPAVSPPQATPTSLQQTPPCSTPPATTNFPAPPTRARLSRPDSGILASRIPLQSYSQVISRGQAPLAPTHTPELSpecifications
NP_071418.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POP2 | |
POPDC2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64091 | |
POPDC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSANSSRVGQLLLQGSACIRWKQDVEGAVYHLANCLLLLGFMGGSGVYGCFYLFGFLSAGYLCCVLWGWFSACGLDIVLWSFLLAVVCLLQLAHLVYRLREDTLPEEFDLLYKTLCLPLQVPLQTYKEIVHCCEEQVLTLATEQTYAVEGETPINRLSLLLSGRVRVSQDGQFLHYIFPYQFMDSPEWESLQPSEEGVFQVTLTAETSCSYISWPRKSLHLLLTKERYISCLFSALLGYDISEKLYTLNDKLFAKFGLRFDIRLPSLYHVLGPTAADAGPESEKGDEEVCEPAVSPPQATPTSLQQTPPCSTPPATTNFPAPPTRARLSRPDSGILASRIPLQSYSQVISRGQAPLAPTHTPEL | |
RUO | |
POPDC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |