missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human POPDC2 Full-length ORF (NP_071418.2, 1 a.a. - 364 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_071418.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64091 |
Molecular Weight (g/mol) | 66.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16143373
|
Abnova™
H00064091-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16133373
|
Abnova™
H00064091-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This gene encodes a member of the POP family of proteins which contain three putative transmembrane domains. This membrane associated protein is predominantly expressed in skeletal and cardiac muscle, and may have an important function in these tissues. [provided by RefSeq]
Sequence: MSANSSRVGQLLLQGSACIRWKQDVEGAVYHLANCLLLLGFMGGSGVYGCFYLFGFLSAGYLCCVLWGWFSACGLDIVLWSFLLAVVCLLQLAHLVYRLREDTLPEEFDLLYKTLCLPLQVPLQTYKEIVHCCEEQVLTLATEQTYAVEGETPINRLSLLLSGRVRVSQDGQFLHYIFPYQFMDSPEWESLQPSEEGVFQVTLTAETSCSYISWPRKSLHLLLTKERYISCLFSALLGYDISEKLYTLNDKLFAKFGLRFDIRLPSLYHVLGPTAADAGPESEKGDEEVCEPAVSPPQATPTSLQQTPPCSTPPATTNFPAPPTRARLSRPDSGILASRIPLQSYSQVISRGQAPLAPTHTPELSpezifikation
NP_071418.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSANSSRVGQLLLQGSACIRWKQDVEGAVYHLANCLLLLGFMGGSGVYGCFYLFGFLSAGYLCCVLWGWFSACGLDIVLWSFLLAVVCLLQLAHLVYRLREDTLPEEFDLLYKTLCLPLQVPLQTYKEIVHCCEEQVLTLATEQTYAVEGETPINRLSLLLSGRVRVSQDGQFLHYIFPYQFMDSPEWESLQPSEEGVFQVTLTAETSCSYISWPRKSLHLLLTKERYISCLFSALLGYDISEKLYTLNDKLFAKFGLRFDIRLPSLYHVLGPTAADAGPESEKGDEEVCEPAVSPPQATPTSLQQTPPCSTPPATTNFPAPPTRARLSRPDSGILASRIPLQSYSQVISRGQAPLAPTHTPEL | |
RUO | |
POPDC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64091 | |
POPDC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POP2 | |
POPDC2 | |
Recombinant | |
wheat germ expression system |