Learn More
Abnova™ Human PRIC285 Partial ORF (NP_208384.2, 1973 a.a. - 2078 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The protein encoded by this gene is a nuclear transcriptional co-activator for peroxisome proliferator activated receptor alpha. The encoded protein contains a zinc finger and is a helicase that appears to be part of the peroxisome proliferator activated receptor alpha interacting complex. This gene is a member of the DNA2/NAM7 helicase gene family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_208384.2 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 85441 |
Molecular Weight (g/mol) | 37.4kDa |
Name | PRIC285 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | GVAVSSITKSQGSEWRYVLVSTVRTCAKSDLDQRPTKSWLKKFLGFVVDPNQVNVAVTRAQEGLCLIGDHLLLRCCPLWRSLLDFCEAQQTLVPAGQVRVCRRPTM |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.