missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Prostaglandin E Synthase 2/PTGES2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
1840.00 SEK - 4470.00 SEK
Specifications
Antigen | Prostaglandin E Synthase 2/PTGES2 |
---|---|
Clone | 998012 |
Dilution | Western Blot 2 ÎĽg/mL, Immunohistochemistry 5-25 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793487
![]() |
R&D Systems
MAB7627-SP |
25 ÎĽg |
1840.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701677
![]() |
R&D Systems
MAB7627-100 |
100 ÎĽg |
4470.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prostaglandin E Synthase 2/PTGES2 Monoclonal specifically detects Prostaglandin E Synthase 2/PTGES2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Prostaglandin E Synthase 2/PTGES2 | |
Western Blot 2 ÎĽg/mL, Immunohistochemistry 5-25 ÎĽg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 ÎĽm filtered solution in PBS. | |
C9orf15, chromosome 9 open reading frame 15, EC 5.3.99.3, FLJ14038, gamma-interferon-activated transcriptional element-binding factor 1, GATE-binding factor 1, GBF1, membrane-associated prostaglandin E synthase 2, Microsomal prostaglandin E synthase 2, microsomal prostaglandin E synthase-2, mPGES2, mPGES-2, PGES2, PGES2MGC11289, prostaglandin E synthase 2 | |
E. coli-derived recombinant Human Prostaglandin E Synthase 2/PTGES2, Glu88-His377, Accession No. Q9H7Z7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Human Prostaglandin E Synthase 2/PTGES2 in direct ELISAs. |
998012 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
Q9H7Z7 | |
80142.0 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Prostaglandin E Synthase 2/PTGES2 Antibody, R&D Systems™