missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human QTRT1 Full-length ORF (AAH15350.1, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
tRNA-guanine transglycosylase (TGT; EC 2.4.2.29) synthesizes queuosine (Q), which is found in tRNAs that recognize NAU and NAC codons, encoding tyr, asn, asp, and his. Prokaryotic TGT is a single protein of 43 kD. In contrast, mammalian TGT appears to be a heterodimer consisting of a 60-kD subunit (USP14; MIM 607274) and a 43-kD catalytic subunit (QTRT1) (Deshpande and Katze, 2001 [PubMed 11255023]).[supplied by OMIM]
Sequence: MPVGTQATMKGITTEQLDALGCRICLGNTYHLGLRPGPELIQKANGLHGFMNWPHNLLTDSGGFQMVSLVSLSEVTEEGVRFRSPYDGNETLLSPEKSVQIQNALGSDIIMQLDDVVSSTVTGPRVEEAMYRSIRWLDRCIAAHQRPDKQNLFAIIQGGLDADLRATCLEEMTKRDVPGFAIGGLSGGESKSQFWRMVALSTSRLPKDKPRYLMGVGYATDLVVCVALGCDMFDCVFPTRTARFGSALVPTGNLQLRKKVFEKDFGPIDPECTCPTCQKHSRAFLHALLHSDNTAALHHLTVHNIAYQLQLMSAVRTSIVEKRFPDFVRDFMGAMYGDPTLCPTWATDALASVGITLG
Specifications
Specifications
Accession Number | AAH15350.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 81890 |
Molecular Weight (g/mol) | 65.7kDa |
Name | QTRT1 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | MPVGTQATMKGITTEQLDALGCRICLGNTYHLGLRPGPELIQKANGLHGFMNWPHNLLTDSGGFQMVSLVSLSEVTEEGVRFRSPYDGNETLLSPEKSVQIQNALGSDIIMQLDDVVSSTVTGPRVEEAMYRSIRWLDRCIAAHQRPDKQNLFAIIQGGLDADLRATCLEEMTKRDVPGFAIGGLSGGESKSQFWRMVALSTSRLPKDKPRYLMGVGYATDLVVCVALGCDMFDCVFPTRTARFGSALVPTGNLQLRKKVFEKDFGPIDPECTCPTCQKHSRAFLHALLHSDNTAALHHLTVHNIAYQLQLMSAVRTSIVEKRFPDFVRDFMGAMYGDPTLCPTWATDALASVGITLG |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human QTRT1 Full-length ORF (AAH15350.1, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction