Learn More
Abnova™ Human RBBP7 Full-length ORF (NP_002884.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005931-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation. [provided by RefSeq]
Sequence: MASKEMFEDTVEERVINEEYKIWKKNTPFLYDLVMTHALQWPSLTVQWLPEVTKPEGKDYALHWLVLGTHTSDEQNHLVVARVHIPNDDAQFDASHCDSDKGEFGGFGSVTGKIECEIKINHEGEVNRARYMPQNPHIIATKTPSSDVLVFDYTKHPAKPDPSGECNPDLRLRGHQKEGYGLSWNSNLSGHLLSASDDHTVCLWDINAGPKEGKIVDAKAIFTGHSAVVEDVAWHLLHESLFGSVADDQKLMIWDTRSNTTSKPSHLVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHTFESHKDEIFQVHWSPHNETILASSGTDRRLNVWDLSKIGEEQSAEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQIWQMAENIYNDEESDVTTSELEGQGSSpecifications
NP_002884.1 | |
Liquid | |
5931 | |
RBBP7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASKEMFEDTVEERVINEEYKIWKKNTPFLYDLVMTHALQWPSLTVQWLPEVTKPEGKDYALHWLVLGTHTSDEQNHLVVARVHIPNDDAQFDASHCDSDKGEFGGFGSVTGKIECEIKINHEGEVNRARYMPQNPHIIATKTPSSDVLVFDYTKHPAKPDPSGECNPDLRLRGHQKEGYGLSWNSNLSGHLLSASDDHTVCLWDINAGPKEGKIVDAKAIFTGHSAVVEDVAWHLLHESLFGSVADDQKLMIWDTRSNTTSKPSHLVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHTFESHKDEIFQVHWSPHNETILASSGTDRRLNVWDLSKIGEEQSAEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQIWQMAENIYNDEESDVTTSELEGQGS | |
RUO | |
RBBP7 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC138867/MGC138868/RbAp46 | |
RBBP7 | |
Yes | |
wheat germ expression system |