Learn More
STEMCELL Technologies Human Recombinant CNTF, ACF
Ciliary neurotrophic factor, animal component-free
Brand:  STEMCELL Technologies 78170.2
Encompass_Preferred
 View more versions of this product
Product Code. 17199741
  • 43426.80 SEK / 1000µg
This item is not returnable. View return policy
Description
Description
  • Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that belongs to the four-helix bundle cytokine family and is structurally related to interleukin 6 (IL-6), interleukin 11 (IL-11), leukemia inhibitory factor (LIF), and oncostatin M (OSM)
  • CNTF binds to its receptor CNFTRα and induces formation of a heterodimer of the signal transducing IL-6 receptor gp130 and LIF receptor (LIFR)-β, which triggers JAK/STAT, ERK, and PI3K signaling cascades
  • CNTF plays an important role in neurogenesis and the differentiation of neural stem cells and has been suggested to possess a therapeutic role in treating neurological disorders
  • Additionally, CNTF is commonly used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into astrocytes
  • This product is animal component-free
Specifications
Specifications
Ciliary neurotrophic factor, HCNTF
1000 μg
Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
≥95%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant CNTF, ACF