Learn More
STEMCELL Technologies Human Recombinant IL-11, ACF
Interleukin 11, animal component-free
Brand:  STEMCELL Technologies 78192
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17199791
  • POA / 10µg
Estimated Shipment: 14-07-2023
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Interleukin 11 (IL-11) is a pleiotropic cytokine with effects on various tissues including the bone marrow, brain, and intestinal mucosa
  • It belongs to the IL-6 family of cytokines that share a common signal transducer, gp130
  • IL-11 induces the proliferation of hematopoietic stem cells and megakaryocytic progenitor cells, the maturation of megakaryocytes, and the production of platelets
  • IL-11 is produced by a variety of cell types including hematopoietic cells, mesenchymal cells, epithelial cells, and neuronal cells
  • It was first cloned from a cDNA library of the human bone marrow-derived stromal cell line KM-102
  • The binding of IL-11 to its receptor induces heterodimerization with the gp130 subunit and activation of JAK tyrosine kinases
  • This product is animal component-free
Specifications
Specifications
Adipogenesis inhibitory factor, AGIF, Interleukin-11, Oprelvekin
≥95%
10 μg
B Cells; Hematopoietic Stem and Progenitor Cells; Other; Pluripotent Stem Cells; T Cells