Learn More
STEMCELL Technologies Human Recombinant IL-5, ACF
Interleukin 5, animal component-free
Brand:  STEMCELL Technologies 78195.2
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17199801
  • POA / 1000µg
Estimated Shipment: 18-07-2023
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Interleukin 5 (IL-5) is a member of the short-chain 4-α-helical bundle subset of hematopoietic cytokines
  • It binds to a receptor consisting of IL-5Ra, which is specific for IL-5R and common beta chain, which is shared with the receptor for IL-3 and granulocyte-macrophage colony-stimulating factor (GM-CSF; Martinez-Moczygemba & Huston)
  • Upon binding to its receptor, IL-5 activates the JAK/STAT and MAPK pathways
  • IL-5 is produced by Th2 cells, eosinophils, and activated mast cells
  • It functions in the recruitment, activation, proliferation, and survival of eosinophils, thus playing an important role in allergic inflammation, asthma, and parasite immunity
  • Stimulation of eosinophils with IL-5 leads to their activation, upregulation of CD11b expression, and inhibition of apoptosis (Martinez-Moczygemba & Huston)
  • This product is animal component-free
Specifications
Specifications
B-cell differentiation factor I, EDF, Eosinophil colony-stimulating factor, Eosinophil differentiation factor, Interleukin-5, T-cell replacing factor, TRF
≥95%
1000 μg
B Cells; Granulocytes and Subsets; Hematopoietic Stem and Progenitor Cells