Learn More
STEMCELL Technologies Human Recombinant IL-5, ACF
Interleukin 5, animal component-free
Brand: STEMCELL Technologies 78195.2
Additional Details : Weight : 0.00110kg
Description
- Interleukin 5 (IL-5) is a member of the short-chain 4-α-helical bundle subset of hematopoietic cytokines
- It binds to a receptor consisting of IL-5Ra, which is specific for IL-5R and common beta chain, which is shared with the receptor for IL-3 and granulocyte-macrophage colony-stimulating factor (GM-CSF; Martinez-Moczygemba & Huston)
- Upon binding to its receptor, IL-5 activates the JAK/STAT and MAPK pathways
- IL-5 is produced by Th2 cells, eosinophils, and activated mast cells
- It functions in the recruitment, activation, proliferation, and survival of eosinophils, thus playing an important role in allergic inflammation, asthma, and parasite immunity
- Stimulation of eosinophils with IL-5 leads to their activation, upregulation of CD11b expression, and inhibition of apoptosis (Martinez-Moczygemba & Huston)
- This product is animal component-free
Specifications
B-cell differentiation factor I, EDF, Eosinophil colony-stimulating factor, Eosinophil differentiation factor, Interleukin-5, T-cell replacing factor, TRF | |
≥95% | |
1000 μg | |
B Cells; Granulocytes and Subsets; Hematopoietic Stem and Progenitor Cells |