missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant TGF-beta 3

Click to view available options
Quantity:
10 μg
100 μg
4 x 250 μg
Unit Size:
100µg
10µg
Pack of 4
Description
- Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes
- TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.) TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.) TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.)
Specifications
Specifications
Name | ARVD, FLJ16571, TGFB3, TGF-b3, Transforming growth factor beta-3 |
Quantity | 100 μg |
Cell Type | Mesenchymal Cells, PSC-Derived; Mesenchymal Stem and Progenitor Cells |
Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant TGF-beta 3
Spot an opportunity for improvement?Share a Content Correction