missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF123 Partial ORF (NP_071347, 1216 a.a. - 1314 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00063891-Q01.25ug

 View more versions of this product

Product Code. 16189466

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. [provided by RefSeq]

Sequence: ADYISADELAQVEQMLAHLTSASAQAAAASLPTSEEDLCPICYAHPISAVFQPCGHKSCKACINQHLMNNKDCFFCKTTIVSVEDWEKGANTSTTSSAA
Specifications

Specifications

NP_071347
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ADYISADELAQVEQMLAHLTSASAQAAAASLPTSEEDLCPICYAHPISAVFQPCGHKSCKACINQHLMNNKDCFFCKTTIVSVEDWEKGANTSTTSSAA
RUO
RNF123
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
63891
RNF123 (Human) Recombinant Protein (Q01)
25 ÎĽg
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686C2222/FLJ12565/FP1477/KPC1/MGC163504
RNF123
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human RNF123 Partial ORF (NP_071347, 1216 a.a. - 1314 a.a.) Recombinant Protein with GST-tag at N-terminal > 25ÎĽg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.