missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human RPL21 Partial ORF (NP_000973, 2 a.a. - 85 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16184575

Abnova™ Human RPL21 Partial ORF (NP_000973, 2 a.a. - 85 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16184575
25 ug, 25µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16184575

Brand: Abnova™ H00006144Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L21E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: TNTKGKRRGTRYMFSRPFRKHGVVPLATYMRIYKKGDIVDIKGMGTVQKGMPHKCYHGKTGRVYNVTQHAVGIVVNKQVKGKIL

Specifications

Accession Number NP_000973
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6144
Molecular Weight (g/mol) 34.98kDa
Name RPL21 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen TNTKGKRRGTRYMFSRPFRKHGVVPLATYMRIYKKGDIVDIKGMGTVQKGMPHKCYHGKTGRVYNVTQHAVGIVVNKQVKGKIL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686C06101/FLJ27458/L21/MGC104274/MGC104275/MGC71252
Common Name RPL21
Gene Symbol RPL21
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human RPL21 Partial ORF (NP_000973, 2 a.a. - 85 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.