missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human RPS6KB1 Partial ORF (NP_003152, 416 a.a. - 525 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16184685

Abnova™ Human RPS6KB1 Partial ORF (NP_003152, 416 a.a. - 525 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16184685
10 ug, 10µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16184685

Brand: Abnova™ H00006198Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding this gene and overexpression of this kinase are seen in some breast cancer cell lines. Alternate translational start sites have been described and alternate transcriptional splice variants have been observed but have not been thoroughly characterized. [provided by RefSeq]

Sequence: PSVLESVKEKFSFEPKIRSPRRFIGSPRTPVSPVKFSPGDFWGRGASASTANPQTPVEYPMETSGIEQMDVTMSGEASAPLPIRQPNSGPYKKQAFPMISKRPEHLRMNL

Specifications

Accession Number NP_003152
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6198
Molecular Weight (g/mol) 37.84kDa
Name RPS6KB1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen PSVLESVKEKFSFEPKIRSPRRFIGSPRTPVSPVKFSPGDFWGRGASASTANPQTPVEYPMETSGIEQMDVTMSGEASAPLPIRQPNSGPYKKQAFPMISKRPEHLRMNL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PS6K/S6K/S6K1/STK14A/p70(S6K)-alpha/p70-S6K/p70-alpha
Common Name RPS6KB1
Gene Symbol RPS6KB1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human RPS6KB1 Partial ORF (NP_003152, 416 a.a. - 525 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.