missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human SALF Partial ORF (NP_758515, 141 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16182777

Abnova™ Human SALF Partial ORF (NP_758515, 141 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16182777
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16182777

Brand: Abnova™ H00286749Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The STON1-GTF2A1L mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring STON1 and GTF2A1L genes. This rare transcript encodes a fusion protein composed of greater than 95% each of the individual elements, stonin 1 and general transcription factor IIA, 1-like. The significance of this co-transcribed mRNA and the function of its protein product have not yet been determined. [provided by RefSeq]

Sequence: SCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLS

Specifications

Accession Number NP_758515
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 286749
Molecular Weight (g/mol) 37.73kDa
Name SALF (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen SCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC126821/MGC126823/SALF
Common Name STON1-GTF2A1L
Gene Symbol STON1-GTF2A1L
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human SALF Partial ORF (NP_758515, 141 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.