missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human SCRG1 Full-length ORF (NP_009212.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16136582

Abnova™ Human SCRG1 Full-length ORF (NP_009212.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16136582
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16136582

Brand: Abnova™ H00011341P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Scrapie-responsive gene 1 is associated with neurodegenerative changes observed in transmissible spongiform encephalopathies. It may play a role in host response to prion-associated infections. The scrapie responsive protein 1 may be partly included in the membrane or secreted by the cells due to its hydrophobic N-terminus. [provided by RefSeq]

Sequence: MKLMVLVFTIGLTLLLGVQAMPANRLSCYRKILKDHNCHNLPEGVADLTQIDVNVQDHFWDGKGCEMICYCNFSELLCCPKDVFFGPKISFVIPCNNQ

Specifications

Accession Number NP_009212.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11341
Molecular Weight (g/mol) 37.5kDa
Name SCRG1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MKLMVLVFTIGLTLLLGVQAMPANRLSCYRKILKDHNCHNLPEGVADLTQIDVNVQDHFWDGKGCEMICYCNFSELLCCPKDVFFGPKISFVIPCNNQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC26468/SCRG-1
Common Name SCRG1
Gene Symbol SCRG1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human SCRG1 Full-length ORF (NP_009212.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.