missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEC24D Partial ORF (NP_055637, 935 a.a. - 1032 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
The protein encoded by this gene is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. This gene product is implicated in the shaping of the vesicle, and also in cargo selection and concentration. [provided by RefSeq]
Sequence: SPPELIQGIFNVPSFAHINTDMTLLPEVGNPYSQQLRMIMGIIQQKRPYSMKLTIVKQREQPEMVFRQFLVEDKGLYGGSSYVDFLCCVHKEICQLLN
Specifications
Specifications
Accession Number | NP_055637 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9871 |
Molecular Weight (g/mol) | 36.41kDa |
Name | SEC24D (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | SPPELIQGIFNVPSFAHINTDMTLLPEVGNPYSQQLRMIMGIIQQKRPYSMKLTIVKQREQPEMVFRQFLVEDKGLYGGSSYVDFLCCVHKEICQLLN |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human SEC24D Partial ORF (NP_055637, 935 a.a. - 1032 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction