missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SEPT12 Full-length ORF (AAH35619.1, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).[supplied by OMIM]
Sequence: MDPLRRSPSPCLSSQPSSPSTPPCEMLGPVGIEAVLDQLKIKAMKMGFEFNIMVVGQSGLGKSTMVNTLFKSKVWKSNPPGLGVPTPQTLQLHSLTHVIEEKGVKLKLTVTDTPGFGDQINNDNCWDPILGYINEQYEQYLQEEILITRQRHIPDTRVHCCVYFVPPTGHCLRPLDIEFLQRLCRTVNVVPVIARADSLTMEEREAFRRRIQQNLRTHCIDVYPQMCFDEDINDKILNSKLRDRIPFAVVGADQEHLVNGRCVLGRKTKWGIIEVENMAHCEFPLLRDLLIRSHLQDLKDITHNIHYENYRVIRLNESHLLPRGPGWVNLAPASPGQLTTPRTFKVCRGAHDDSDDEF
Specifications
Specifications
Accession Number | AAH35619.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 124404 |
Molecular Weight (g/mol) | 67.1kDa |
Name | SEPT12 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | MDPLRRSPSPCLSSQPSSPSTPPCEMLGPVGIEAVLDQLKIKAMKMGFEFNIMVVGQSGLGKSTMVNTLFKSKVWKSNPPGLGVPTPQTLQLHSLTHVIEEKGVKLKLTVTDTPGFGDQINNDNCWDPILGYINEQYEQYLQEEILITRQRHIPDTRVHCCVYFVPPTGHCLRPLDIEFLQRLCRTVNVVPVIARADSLTMEEREAFRRRIQQNLRTHCIDVYPQMCFDEDINDKILNSKLRDRIPFAVVGADQEHLVNGRCVLGRKTKWGIIEVENMAHCEFPLLRDLLIRSHLQDLKDITHNIHYENYRVIRLNESHLLPRGPGWVNLAPASPGQLTTPRTFKVCRGAHDDSDDEF |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human SEPT12 Full-length ORF (AAH35619.1, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction