missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SIN3B Partial ORF (NP_056075.1, 1063 a.a. - 1160 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4630.00 SEK - 7020.00 SEK

Specifications

Accession Number NP_056075.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23309
Molecular Weight (g/mol) 36.52kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16194556
View Documents
Abnova™
H00023309-Q01.10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16104566
View Documents
Abnova™
H00023309-Q01.25ug
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Sequence: HKMVFIVNSEDYMYRRGTLCRAKQVQPLVLLRHHQHFEEWHSRWLEDNVTVEAASLVQDWLMGEEDEDMVPCKTLCETVHVHGLPVTRYRVQYSRRPA
Specifications

Specifications

NP_056075.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0700
SIN3B
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
23309
SIN3B (Human) Recombinant Protein (Q01)
HKMVFIVNSEDYMYRRGTLCRAKQVQPLVLLRHHQHFEEWHSRWLEDNVTVEAASLVQDWLMGEEDEDMVPCKTLCETVHVHGLPVTRYRVQYSRRPA
RUO
SIN3B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human SIN3B Partial ORF (NP_056075.1, 1063 a.a. - 1160 a.a.) Recombinant Protein with GST-tag at N-terminal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.