Learn More
Abnova™ Human SIPA1 Partial ORF (AAH10492, 933 a.a. - 1042 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date. [provided by RefSeq]
Specifications
Specifications
Accession Number | AAH10492 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6494 |
Molecular Weight (g/mol) | 37.73kDa |
Name | SIPA1 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 ug |
Immunogen | SEIASTCNTILESLSREGQPIPESGDPKGTPKSDAEPEPGNLSEKVSHLESMLRKLQEDLQKEKADRAALEEEVRSLRHNNRRLQAESESAATRLLLASKQLGSPTADLA |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.