missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human SLC25A40 Full-length ORF (NP_061331.2, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16162003

Abnova™ Human SLC25A40 Full-length ORF (NP_061331.2, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16162003
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16162003

Brand: Abnova™ H00055972P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

SLC25A40 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]

Sequence: MDPETRGQEIIKVTPLQQMLASCTGAILTSVIVTPLDVVKIRLQAQNNPLPKGKCFVYSNGLMDHLCVCEEGGNKLWYKKPGNFQGTLDAFFKIIRNEGIKSLWSGLPPTLVMAVPATVIYFTCYDQLSALLRSKLGENETCIPIVAGIVARFGAVTVISPLELIRTKMQSKKFSYVELHRFVSKKVSEDGWISLWRGWAPTVLRDVPFSAMYWYNYEILKKWLCEKSGLYEPTFMINFTSGALSGSFAAVATLPFDVVKTQKQTQLWTYESHKISMPLHMSTWIIMKNIVAKNGFSGLFSGLIPRLIKIAPACAIMISTYEFGKAFFQKQNVRRQQY

Specifications

Accession Number NP_061331.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55972
Molecular Weight (g/mol) 64.5kDa
Name SLC25A40 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MDPETRGQEIIKVTPLQQMLASCTGAILTSVIVTPLDVVKIRLQAQNNPLPKGKCFVYSNGLMDHLCVCEEGGNKLWYKKPGNFQGTLDAFFKIIRNEGIKSLWSGLPPTLVMAVPATVIYFTCYDQLSALLRSKLGENETCIPIVAGIVARFGAVTVISPLELIRTKMQSKKFSYVELHRFVSKKVSEDGWISLWRGWAPTVLRDVPFSAMYWYNYEILKKWLCEKSGLYEPTFMINFTSGALSGSFAAVATLPFDVVKTQKQTQLWTYESHKISMPLHMSTWIIMKNIVAKNGFSGLFSGLIPRLIKIAPACAIMISTYEFGKAFFQKQNVRRQQY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MCFP
Common Name SLC25A40
Gene Symbol SLC25A40
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human SLC25A40 Full-length ORF (NP_061331.2, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.