Learn More
Invitrogen™ Human SLC4A7 (aa 240-319) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP96255
Description
Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57433 (PA5-57433. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
Specifications
Q9Y6M7 | |
Blocking Assay, Control | |
9497 | |
100 ÎĽL | |
bicarbonate transporter; BT; E430014N10Rik; Electroneutral Na/HCO(3) cotransporter; electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; Nbc; NBC2; NBC2B; NBC3; NBC-like protein; Nbcn1; NBCn1-G; SBC2; SLC4A6; Slc4a7; Sodium bicarbonate cotransporter 2; sodium bicarbonate cotransporter 2 b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7 | |
SLC4A7 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human SLC4A7 (aa 240-319) Control Fragment | |
RUO | |
SLC4A7 | |
Unconjugated | |
Recombinant | |
KHSDPHLLERNGEGLSASRHSLRTGLSASNLSLRGESPLSLLLGHLLPSSRAGTPAGSRCTTPVPTPQNSPPSSPSISRL | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.