missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC7A8 Partial ORF (NP_036376.2, 467 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16184666
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: VYWQHKPKCFSDFIELLTLVSQKMCVVVYPEVERGSGTEEANEDMEEQQQPMYQPTPTKDKDVAGQPQP

Specifications

Accession Number NP_036376.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23428
Molecular Weight (g/mol) 33.33kDa
Name SLC7A8 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen VYWQHKPKCFSDFIELLTLVSQKMCVVVYPEVERGSGTEEANEDMEEQQQPMYQPTPTKDKDVAGQPQP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LAT2/LPI-PC1
Common Name SLC7A8
Gene Symbol SLC7A8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human SLC7A8 Partial ORF (NP_036376.2, 467 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.