Learn More
Abnova™ Human SP3 Partial ORF (NP_003102.1, 516 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006670-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. [provided by RefSeq]
Sequence: GQLPNLQTVTVNSIDSAGIQLHPGENADSPADIRIKEEEPDPEEWQLSGDSTLNTNDLTHLRVQVVDEEGDQQHQEGKRLRRVACTCPNCKEGGGRGTNLSpecifications
NP_003102.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GQLPNLQTVTVNSIDSAGIQLHPGENADSPADIRIKEEEPDPEEWQLSGDSTLNTNDLTHLRVQVVDEEGDQQHQEGKRLRRVACTCPNCKEGGGRGTNL | |
RUO | |
SP3 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
6670 | |
SP3 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686O1631|SPR-2 | |
SP3 | |
Yes |