Learn More
Abnova™ Human STCH Full-length ORF (NP_008879.3, 1 a.a. - 471 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006782-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein. [provided by RefSeq]
Sequence: MAREMTILGSAVLTLLLAGYLAQQYLPLPTPKVIGIDLGTTYCSVGVFFPGTGKVKVIPDENGHISIPSMVSFTDNDVYVGYESVELADSNPQNTIYDAKRFIGKIFTAEELEAEIGRYPFKVLNKNGMVEFSVTSNETITVSPEYVGSRLLLKLKEMAEAYLGMPVANAVISVPAEFDLKQRNSTIEAANLAGLKILRVINEPTAAAMAYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDHRVNSGFGRGLSDKKSGESQVLFETEISRKLFDTLNEDLFQKILVPIQQVLKEGHLEKTEIDEVVLVGGSTRIPRIRQVIQEFFGKDPNTSVDPDLAVVTGVAIQAGIDGGSWPLQVSALEIPNKHLQKTNFNSpecifications
NP_008879.3 | |
Liquid | |
6782 | |
STCH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAREMTILGSAVLTLLLAGYLAQQYLPLPTPKVIGIDLGTTYCSVGVFFPGTGKVKVIPDENGHISIPSMVSFTDNDVYVGYESVELADSNPQNTIYDAKRFIGKIFTAEELEAEIGRYPFKVLNKNGMVEFSVTSNETITVSPEYVGSRLLLKLKEMAEAYLGMPVANAVISVPAEFDLKQRNSTIEAANLAGLKILRVINEPTAAAMAYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDHRVNSGFGRGLSDKKSGESQVLFETEISRKLFDTLNEDLFQKILVPIQQVLKEGHLEKTEIDEVVLVGGSTRIPRIRQVIQEFFGKDPNTSVDPDLAVVTGVAIQAGIDGGSWPLQVSALEIPNKHLQKTNFN | |
RUO | |
HSPA13 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
STCH | |
HSPA13 | |
Yes | |
wheat germ expression system |