missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human TAS1R1 Partial ORF (NP_619642, 281 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16185053

Abnova™ Human TAS1R1 Partial ORF (NP_619642, 281 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16185053
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16185053

Brand: Abnova™ H00080835Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a G protein-coupled receptor and is a component of the heterodimeric amino acid taste receptor T1R1+3. The T1R1+3 receptor responds to L-amino acids but not to D-enantiomers or other compounds. Most amino acids that are perceived as sweet activate T1R1+3, and this activation is strictly dependent on an intact T1R1+3 heterodimer. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]

Sequence: RVFFESVVLTNLTGKVWVASEAWALSRHITGVPGIQRIGMVLGVAIQKRAVPGLKAFEEAYARADKEAPRPCHKGSWCSSNQLCRECQAFMAHTMPKLKA

Specifications

Accession Number NP_619642
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 80835
Molecular Weight (g/mol) 36.74kDa
Name TAS1R1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen RVFFESVVLTNLTGKVWVASEAWALSRHITGVPGIQRIGMVLGVAIQKRAVPGLKAFEEAYARADKEAPRPCHKGSWCSSNQLCRECQAFMAHTMPKLKA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GPR70/T1R1/TR1/gm148
Common Name TAS1R1
Gene Symbol TAS1R1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human TAS1R1 Partial ORF (NP_619642, 281 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.