Learn More
Abnova™ Human TESK2 Partial ORF (AAH33085, 405 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene product is a serine/threonine protein kinase that contains an N-terminal protein kinase domain that is structurally similar to the kinase domains of testis-specific protein kinase-1 and the LIM motif-containing protein kinases (LIMKs). Its overall structure is most related to the former, indicating that it belongs to the TESK subgroup of the LIMK/TESK family of protein kinases. This gene is predominantly expressed in testis and prostate. The developmental expression pattern of the rat gene in testis suggests an important role for this gene in meitoic stages and/or early stages of spermiogenesis. [provided by RefSeq]
Specifications
Specifications
Accession Number | AAH33085 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10420 |
Molecular Weight (g/mol) | 40.81kDa |
Name | TESK2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | GPGTMPLADWQEPLAPPIRRWCSLPGSPEFLHQEACPFVGREESLSDGPPPRLSSLKYRVKEIPPFRASALPAAQAHEAMDCSILQEENGFGSRPQGTSPCPAGASEEMEVEERPAGSTPATFSTSGIGLQTQGKQDG |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.