Learn More
Abnova™ Human TM4SF1 Full-length ORF (NP_055035.1, 1 a.a. - 202 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface antigen and is highly expressed in different carcinomas. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_055035.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4071 |
Molecular Weight (g/mol) | 48kDa |
Name | TM4SF1 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 ug |
Immunogen | MCYGKCARCIGHSLVGLALLCIAANILLYFPNGETKYASENHLSRFVWFFSGIVGGGLLMLLPAFVFIGLEQDDCCGCCGHENCGKRCAMLSSVLAALIGIAGSGYCVIVAALGLAEGPLCLDSLGQWNYTFASTEGQYLLDTSTWSECTEPKHIVEWNVSLFSILLALGGIEFILCLIQVINGVLGGICGFCCSHQQQYDC |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.