missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human TRAF4 Partial ORF (AAH01769, 371 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16181046

Abnova™ Human TRAF4 Partial ORF (AAH01769, 371 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16181046
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16181046

Brand: Abnova™ H00009618Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the TNF receptor associated factor (TRAF) family. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. The encoded protein has been shown to interact with neurotrophin receptor, p75 (NTR/NTSR1), and negatively regulate NTR induced cell death and NF-kappa B activation. This protein has been found to bind to p47phox, a cytosolic regulatory factor included in a multi-protein complex known as NAD(P)H oxidase. This protein thus, is thought to be involved in the oxidative activation of MAPK8/JNK. Alternatively spliced transcript variants have been observed but the full-length nature of only one has been determined. [provided by RefSeq]

Sequence: PGAFDNLLEWPFARRVTFSLLDQSDPGLAKPQHVTETFHPDPNWKNFQKPGTWRGSLDESSLGFGYPKFISHQDIRKRNYVRDDAVFIRAAVELPRKILS

Specifications

Accession Number AAH01769
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9618
Molecular Weight (g/mol) 36.63kDa
Name TRAF4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PGAFDNLLEWPFARRVTFSLLDQSDPGLAKPQHVTETFHPDPNWKNFQKPGTWRGSLDESSLGFGYPKFISHQDIRKRNYVRDDAVFIRAAVELPRKILS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CART1/MLN62/RNF83
Common Name TRAF4
Gene Symbol TRAF4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human TRAF4 Partial ORF (AAH01769, 371 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.