missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRAF5 Full-length ORF (NP_001029082.1, 1 a.a. - 557 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16183798
10 ug, 10µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The scaffold protein encoded by this gene is a member of the tumor necrosis factor receptor-associated factor (TRAF) protein family and contains a meprin and TRAF homology (MATH) domain, a RING-type zinc finger, and two TRAF-type zinc fingers. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. This protein is one of the components of a multiple protein complex which binds to tumor necrosis factor (TNF) receptor cytoplasmic domains and mediates TNF-induced activation. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]

Sequence: MAYSEEHKGMPCGFIRQNSGNSISLDFEPSIEYQFVERLEERYKCAFCHSVLHNPHQTGCGHRFCQHCILSLRELNTVPICPVDKEVIKSQEVFKDNCCKREVLNLYVYCSNAPGCNAKVILGRYQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKRRNLQQHEHSALREHMRLVLEKNVQLEEQISDLHKSLEQKESKIQQLAETIKKLEKEFKQFAQLFGKNGSFLPNIQVFASHIDKSAWLEAQVHQLLQMVNQQQNKFDLRPLMEAVDTVKQKITLLENNDQRLAVLEEETNKHDTHINIHKAQLSKNEERFKLLEGTCYNGKLIWKVTDYKMKKREAVDGHTVSIFSQSFYTSRCGYRLCARAYLNGDGSGRGSHLSLYFVVMRGEFDSLLQWPFRQRVTLMLLDQSGKKNIMETFKPDPNSSSFKRPDGEMNIASGCPRFVAHSVLENAKNAYIKDDTLFLKVAVDLTDLEDL

Specifications

Accession Number NP_001029082.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7188
Molecular Weight (g/mol) 90.8kDa
Name TRAF5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MAYSEEHKGMPCGFIRQNSGNSISLDFEPSIEYQFVERLEERYKCAFCHSVLHNPHQTGCGHRFCQHCILSLRELNTVPICPVDKEVIKSQEVFKDNCCKREVLNLYVYCSNAPGCNAKVILGRYQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKRRNLQQHEHSALREHMRLVLEKNVQLEEQISDLHKSLEQKESKIQQLAETIKKLEKEFKQFAQLFGKNGSFLPNIQVFASHIDKSAWLEAQVHQLLQMVNQQQNKFDLRPLMEAVDTVKQKITLLENNDQRLAVLEEETNKHDTHINIHKAQLSKNEERFKLLEGTCYNGKLIWKVTDYKMKKREAVDGHTVSIFSQSFYTSRCGYRLCARAYLNGDGSGRGSHLSLYFVVMRGEFDSLLQWPFRQRVTLMLLDQSGKKNIMETFKPDPNSSSFKRPDGEMNIASGCPRFVAHSVLENAKNAYIKDDTLFLKVAVDLTDLEDL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC:39780/RNF84
Common Name TRAF5
Gene Symbol TRAF5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human TRAF5 Full-length ORF (NP_001029082.1, 1 a.a. - 557 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.