Learn More
Abnova™ Human TRIM6-TRIM34 Partial ORF (NP_001003819, 641 a.a. - 740 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This gene represents a read-through transcript transcribed from genes TRIM6 and TRIM34, and it was described as a splice variant of TRIM34. This gene is mapped to chromosome 11p15, where it resides within a TRIM gene cluster. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_001003819 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 445372 |
Molecular Weight (g/mol) | 36.74kDa |
Name | TRIM6-TRIM34 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | LQMFRELTAVRCYWVDVTLNSVNLNLNLVLSEDQRQVISVPIWPFQCYNYGVLGSQYFSSGKHYWEVDVSKKTAWILGVYCRTYSRHMKYVVRRCANRQN |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.