missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TSC22D1 Full-length ORF (AAH16867, 1 a.a. - 144 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008848-P02.25ug
Additional Details : Weight : 0.00010kg
Description
TSC22D1 encodes a transcription factor and belongs to the large family of early response genes.[supplied by OMIM]
Sequence: MKSQWCRPVAMDLGVYQLRHFSISFLSSLLGTENASVRLDNSSSGASVVAIDNKIEQAMDLVKSHLMYAVREEVEVLKEQIKELIEKNSQLEQENNLLKTLASPEQLAQFQAQLQTGSPPATTQPQGTTQPPAQPASQGSGPTASpecifications
AAH16867 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
41.58kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686O19206/MGC17597/RP11-269C23.2/TGFB1I4/TSC22 | |
TSC22D1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8848 | |
TSC22D1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKSQWCRPVAMDLGVYQLRHFSISFLSSLLGTENASVRLDNSSSGASVVAIDNKIEQAMDLVKSHLMYAVREEVEVLKEQIKELIEKNSQLEQENNLLKTLASPEQLAQFQAQLQTGSPPATTQPQGTTQPPAQPASQGSGPTA | |
RUO | |
TSC22D1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |