missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human TSPAN2 Partial ORF (NP_005716, 112 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16181846

Abnova™ Human TSPAN2 Partial ORF (NP_005716, 112 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16181846
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16181846

Brand: Abnova™ H00010100Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. [provided by RefSeq]

Sequence: GKGVAIRHVQTMYEEAYNDYLKDRGKGNGTLITFHSTFQCCGKESSEQVQPTCPKELLGHKNCIDEIETIISVKLQ

Specifications

Accession Number NP_005716
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10100
Molecular Weight (g/mol) 34.1kDa
Name TSPAN2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen GKGVAIRHVQTMYEEAYNDYLKDRGKGNGTLITFHSTFQCCGKESSEQVQPTCPKELLGHKNCIDEIETIISVKLQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 6330415F13Rik/FLJ12082/TSN2/TSPAN-2
Common Name TSPAN2
Gene Symbol TSPAN2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human TSPAN2 Partial ORF (NP_005716, 112 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.