Learn More
Abnova™ Human UBXD5 Partial ORF (NP_663320.1, 311 a.a. - 409 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_663320.1 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 91544 |
Molecular Weight (g/mol) | 36.41kDa |
Name | UBXD5 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | QGEVIDIRGPIRDTLQNCCPLPARIQEIVVETPTLAAERERSQESPNTPAPPLSMLRIKSENGEQAFLLMMQPDNTIGDVRALLAQARVMDASAFEIFS |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.