missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human VISTA/B7-H5/PD-1H Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB71266-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifikationer
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession # Q9H7M9 | |
| 100 μg | |
| Primary | |
| Detects human VISTA/B7-H5/PD-1H in direct ELISAs. | |
| Human | |
| Purified |
| Blocking Assay | |
| 1011478 | |
| Blockade of Receptor-ligand Interaction | |
| Q9H7M9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64115 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering