missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VISTA/B7-H5/PD-1H Antibody, R&D Systems™

Mouse Monoclonal Antibody
1925.00 SEK - 4550.00 SEK
Spezifikation
| Antigen | VISTA/B7-H5/PD-1H |
|---|---|
| Klona | 1011478 |
| Utspädning | Blockade of Receptor-ligand Interaction |
| Användningsområden | Blocking Assay |
| Klassificering | Monoclonal |
| Produktcode | Marke | Kvantitet | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Kvantitet | Preis | Menge & Verfügbarkeit | |||||
18731357
![]() |
R&D Systems
MAB71266-SP |
25 μg |
1925.00 SEK
25µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
18723367
![]() |
R&D Systems
MAB71266-100 |
100 μg |
4550.00 SEK
100µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beschreibung
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Spezifikation
| VISTA/B7-H5/PD-1H | |
| Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession # Q9H7M9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VISTA/B7-H5/PD-1H in direct ELISAs. |
| 1011478 | |
| Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H7M9 | |
| 64115 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts