Learn More
Invitrogen™ Human VPS26A (aa 302-327) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP103683
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84347 (PA5-84347. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
O75436 | |
Blocking Assay, Control | |
9559 | |
100 ÎĽL | |
AA407240; FLJ12930; H beta 58; HB58; Hbeta58; Hbeta58 homolog; hVPS26; mVPS26; OTTHUMP00000019721; PEP8A; vacuolar protein sorting 26 A; vacuolar protein sorting 26 homolog A; vacuolar protein sorting 26 homolog A (S. pombe); vacuolar protein sorting 26 homolog A (yeast); vacuolar protein sorting-associated protein 26 A; vacuole protein sorting 26; vesicle protein sorting 26 A; Vps26; VPS26 retromer complex comonent A; VPS26 retromer complex component A; VPS26, retromer complex component A; VPS26A | |
Vps26a | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human VPS26A (aa 302-327) Control Fragment | |
RUO | |
VPS26A | |
Unconjugated | |
Recombinant | |
LRKQRTNFHQRFESPESQASAEQPEM | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.